Learn More
CD63 Monoclonal Antibody (H5C6), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£104.00 - £265.00
Specifications
Antigen | CD63 |
---|---|
Clone | H5C6 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15599686
|
Affymetrix eBioscience
48-0639-42 |
100 Tests |
£265.00
100 tests |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15589686
|
Affymetrix eBioscience
48-0639-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This H5C6 monoclonal antibody reacts with human CD63, a type III member of the tetraspanin family of transmembrane proteins. CD63 is expressed intracellularly on lysosomes, endosomes, and granules of resting platelets and basophils. However, cell surface expression of CD63 can be detected on activated basophils and platelets, monocytes, macrophages, and granulocytes. This receptor is also expressed on endothelial cells, fibroblasts, and smooth muscle cells. Studies have demonstrated that CD63 associates with integrins (VLA-3 and VLA-6) and TIMP-1 to mediate the allergic response. Applications Reported: This H5C6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H5C6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Flow Cytometry.Specifications
CD63 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P08962 | |
CD63 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD63 |
H5C6 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
967 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.