missing translation for 'onlineSavingsMsg'
Learn More

CD62P (P-Selectin), PE, clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0628-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15526806

  • £61.93 / 25 tests


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AK-4 monoclonal antibody reacts with human CD62P, a 140kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils.Applications Reported: The AK-4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This AK-4 antibody has been pre-titrated and tested by flow cytometric analysis of resting and thrombin-activated human platelets. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD62P (P-Selectin)
AK-4
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LECAM-3
IgG1
RUO
Primary
Human
Videos