missing translation for 'onlineSavingsMsg'
Learn More

CD62P (P-Selectin) Mouse anti-Human, FITC, Clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0628-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596426

  • £52.50 / 25 tests

Estimated Shipment
06-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

P-selectin is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate.
Specifications

Specifications

CD62P (P-Selectin)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P16109
RP1-86F14.2, CD62, CD62P, FLJ45155, GMP140, GRMP, LECAM3, PADGEM, PSEL
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
AK-4
FITC
SELP
Liquid
SELP
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6403
Videos
Documents

For Research Use Only.