missing translation for 'onlineSavingsMsg'
Learn More

CD62P (P-Selectin), clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0628-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15217047

  • £31.05 / 25µg


Estimated Shipment
28-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AK-4 monoclonal antibody reacts with human CD62P, a 140kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils.Applications Reported: The AK-4 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. AK-4 has also been reported in blocking adhesion of platelets in in vitro assays. (Please use Functional Grade purified AK-4, cat. 16-0628, in functional assays.).Applications Tested: This AK-4 antibody has been tested by flow cytometric analysis of resting and thrombin-activated human platelets. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD62P (P-Selectin)
AK-4
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry (Frozen)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LECAM-3
IgG1
RUO
Primary
Human
Videos