missing translation for 'onlineSavingsMsg'
Learn More

CD62L (L-Selectin) Mouse anti-Human, APC, Clone: DREG-56 (DREG56), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0629-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15567956

  • £84.75 / 25 tests

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD62L (L-selectin) is an adhesion glycoprotein that is constitutively expressed on the cell surface of leukocytes and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similarly to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1).
Specifications

Specifications

CD62L (L-selectin)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P14151
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DREG-56 (DREG56)
APC
SELL
Liquid
SELL
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
6402
Videos
Documents

For Research Use Only.