Learn More
CD61 (Integrin beta 3) Monoclonal Antibody (VI-PL2), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 11-0619-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The VI-PL2 monoclonal antibody reacts with human CD61, also known as integrin beta 3 and GPIIIa. CD61 is a 90-110 kDa member of the beta integrin family expressed by a wide variety of cells, including leukocytes, platelets, endothelial and smooth muscle cells. CD61 binds non-covalently with the alpha integrins CD41 and CD51, to form the alpha IIb beta3 (CD41/CD61) and alpha v beta3 (CD51/CD61) complexes. These alpha beta heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. The CD41/CD61 integrin has been shown to be involved in platelet aggregation, and binds to fibrinogen, von Willebrand Factor (vWF) and fibronectin. The CD51/CD61 integrin binds to matrix proteins including vitronectin, fibronectin, vWF and fibrinogen, and has been shown to have a strong role in modulating the migration and survival of angiogenic endothelial cells. This VI-PL2 antibody has also been shown to crossreact with canine and several non-human primate species, including baboon, rhesus, and cynomologus monkey. Applications Reported: This VI-PL2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This VI-PL2 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (025 μg) per test.
A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. The ITGB3 protein product is the integrin beta chain beta 3. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. Integrin beta 3 is found along with the alpha IIb chain in platelets. Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling.Specifications
CD61 (Integrin beta 3) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P05106 | |
ITGB3 | |
Affinity chromatography | |
RUO | |
102117791, 3690, 403788, 717379 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
VI-PL2 | |
FITC | |
ITGB3 | |
BDPLT2, CD61, GP3A, GPIIIa, GT, integrin beta-3, platelet glycoprotein IIIa, platelet membrane glycoprotein IIIa, ITGB3 | |
Mouse | |
25 Tests | |
Primary | |
Baboon, Canine, Cynomolgus Monkey, Human, Rhesus Monkey | |
Antibody | |
IgG1 κ |
For Research Use Only.