Learn More
CD57 Monoclonal Antibody (TB01 (TBO1)), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-0577-42
166.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110-kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells. Applications Reported: This TB01 (TBO1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This TB01 (TBO1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1.0 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Flow Cytometry.Specifications
CD57 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9P2W7 | |
B3GAT1 | |
Affinity chromatography | |
RUO | |
27087 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TB01 (TBO1) | |
PE-Cyanine7 | |
B3GAT1 | |
HNK-1; Leu-7 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |
For Research Use Only.