Learn More
CD57 Monoclonal Antibody (TB01 (TBO1)), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£72.50 - £211.00
Specifications
Antigen | CD57 |
---|---|
Clone | TB01 (TBO1) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15566796
|
Affymetrix eBioscience
12-0577-42 |
100 Tests |
£211.00
100 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15556796
|
Affymetrix eBioscience
12-0577-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110-kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells. Applications Reported: This TB01 (TBO1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This TB01 (TBO1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD57, also known as HNK-1 and Leu-7, is an oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitin sulphate proteoglycans. Its function is poorly understood. CD57 is expressed on a subset of natural killer (NK) cells and NK T cells.Specifications
CD57 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9P2W7 | |
B3GAT1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
B3GAT1 |
TB01 (TBO1) | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
27087 | |
IgM | |
Affinity chromatography | |
Antibody |
For Research Use Only.