missing translation for 'onlineSavingsMsg'
Learn More

CD57, eFluor 660, clone: TB01 (TBO1), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-0577-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15509886

  • £68.96 / 25 tests


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Description

Description

Description: This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110-kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells.Applications Reported: This TB01 (TBO1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This TB01 (TBO1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD57
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9P2W7
Cluster designation 57
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TB01 (TBO1)
eFluor 660
B3GAT1
Conjugated
B3GAT1
IgM
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
27087
Videos

For Research Use Only.