missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57, eFluor 450, clone: TB01 (TBO1), eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 48-0577-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110-kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells.Applications Reported: This TB01 (TBO1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This TB01 (TBO1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD57 | |
Monoclonal | |
5μl/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9P2W7 | |
Cluster designation 57 | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
TB01 (TBO1) | |
eFluor 450 | |
B3GAT1 | |
Conjugated | |
B3GAT1 | |
IgM | |
25 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
27087 |
For Research Use Only.