Learn More
CD56 (NCAM) Monoclonal Antibody (TULY56), Super Bright™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 78-0566-41
98.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Description: The MEL-14 monoclonal antibody reacts with mouse CD62L, a 76 kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. Applications Reported: This MEL-14 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MEL-14 antibody has been tested by flow cytomeric analysis of mouse splenocytes. This can be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 780 nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD56 (NCAM) Monoclonal antibody specifically detects CD56 (NCAM) in Human, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD56 (NCAM) | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P13591 | |
Ncam1 | |
Affinity chromatography | |
RUO | |
4684, 693789 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TULY56 | |
Super Bright 780 | |
Ncam1 | |
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
Mouse | |
25 Tests | |
Primary | |
Human, Rhesus Monkey | |
Antibody | |
IgG1 κ |