missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 / NCAM Mouse, PE-Cy7, Clone: MY31, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB48-PE7-25Tests
Additional Details : Weight : 0.01000kg
Specifications
CD56 / NCAM | |
MY31 | |
PE-Cyanine7 | |
NP_000606 | |
Mouse | |
Protein A/G Purified | |
RUO | |
Primary | |
4684 |
Flow Cytometry | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
NCAM1 | |
IgG1 kappa | |
25 Tests | |
Store at 2-4°C. | |
Monoclonal |