missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM) Mouse, eFluor 506, Clone: TULY56, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 69-0566-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510677

  • £72.77 / 25 tests


Estimated Shipment date
from Supplier 01-09-2017
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB.

The TULY56 monoclonal antibody crossreacts with Rhesus macaque.
Description & Specifications

Specifications

CD56 (NCAM)
TULY56
eFluor 506
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Neural cell adhesion molecule
IgG1
RUO
Primary
Rhesus