missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), eFluor 506, clone: TULY56, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 69-0566-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15520677

  • £149.92 / 100 tests


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB.The TULY56 monoclonal antibody crossreacts with Rhesus macaque.Applications Reported: This TULY56 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TULY56 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56 (NCAM)
TULY56
eFluor 506
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Neural cell adhesion molecule
IgG1
RUO
Primary
Rhesus
Videos