Learn More
CD56 (NCAM) Monoclonal Antibody (CMSSB), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£87.50 - £234.00
Specifications
Antigen | CD56 (NCAM) |
---|---|
Clone | CMSSB |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-eFluor 610 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570447
|
Affymetrix eBioscience
61-0567-42 |
100 Tests |
£234.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15560447
|
Affymetrix eBioscience
61-0567-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56. Applications Reported: This CMSSB antibody has been reported for use in flow cytometric analysis. Applications Tested: This CMSSB antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD56 (NCAM) Monoclonal antibody specifically detects CD56 (NCAM) in Human samples. It is validated for Flow Cytometry.Specifications
CD56 (NCAM) | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human | |
P13591 | |
IgG1 κ | |
Affinity chromatography |
CMSSB | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
4684 | |
Primary | |
Antibody |
For Research Use Only.