CD56 (NCAM) Mouse anti-Human, eFluor 660, Clone: 5tukon56, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 50-0565-82

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15599876

Quantity Price
1 £ 155.16 / 100µg
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Specification

Clone 5tukon56
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Regulatory Status RUO
Antigen CD56 (NCAM)
Conjugate eFluor 660
Primary or Secondary Primary
Format Conjugated
Isotype IgG1
Concentration 0.2mg/mL
Quantity 100μg
Monoclonal or Polyclonal Monoclonal
Gene Alias Neural cell adhesion molecule
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Species Reactivity Human
Host Species Mouse

This 5tukon56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed in both the developing and adult nervous system and plays a role in neurogenesis, migration, neurite extension, and the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.

For flow cytometric analysis of human CD56, we recommend clones CMSSB or TULY56.