missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), eFluor 450, clone: TULY56, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0566-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15529686

  • £74.54 / 25 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB.The TULY56 monoclonal antibody crossreacts with Rhesus macaque.Applications Reported: This TULY56 antibody has been reported for use in flow cytometric analysis.Applications Tested: This TULY56 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56 (NCAM)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
HumanþMacaqueþMonkeyþNon-human PrimateþRhesus
Flow Cytometry
TULY56
eFluor 450
NCAM1
Conjugated
NCAM1
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
4684
Videos

For Research Use Only.