missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0567-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15277027

  • £43.32 / 25µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.Applications Reported: This CMSSB antibody has been reported for use in flow cytometric analysis and immunohistology staining of paraffin embedded tissue sections.Applications Tested: This CMSSB antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56 (NCAM)
CMSSB
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry (Formalin/Paraffin)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Neural cell adhesion molecule
IgG1
RUO
Primary
Human
Videos