missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), clone: 5tukon56, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0565-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15597416

  • £68.75 / 25µg

Estimated Shipment
03-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This 5tuckon56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed in both the developing and adult nervous system and plays a role in neurogenesis, migration, neurite extension, and the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells.For flow cytometric analysis of human CD56, we recommend clones CMSSB or TULY56.Applications Reported: This 5tukon56 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy.Applications Tested: This 5tukon56 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue sections using low or high pH antigen retreival and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56 (NCAM)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
5tukon56
Unconjugated
NCAM1
Liquid
NCAM1
IgG1, kappa, kappa
25μg
Store at 2-8°C.
Primary
4684
Videos
SDS
Documents

For Research Use Only.