missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), Biotin, clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0567-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229609

  • £63.31 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.Applications Reported: This CMSSB antibody has been reported for use in flow cytometric analysis and immunohistology staining of paraffin embedded tissue sections.Applications Tested: This CMSSB antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56 (NCAM)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry
CMSSB
Biotin
NCAM1
Conjugated
NCAM1
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
4684
Videos

For Research Use Only.