missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM) Mouse anti-Human, APC, Clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0567-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15587946

  • £71.50 / 25 tests

Estimated Shipment date
from Supplier 09-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.
Specifications

Specifications

CD56 (NCAM)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CMSSB
APC
NCAM1
Liquid
NCAM1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
4684
Videos
Documents

For Research Use Only.