missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56/NCAM-1 Ab-4 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD56/NCAM-1 Ab-4, Mouse Monoclonal Antibody.
£98.23 - £490.00
Specifications
Antigen | CD56 Ab-4 |
---|---|
Clone | 56C04; same as 123A8 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12699096
|
Epredia
MS-1149-P1 |
Concentrated | 500 μL |
£320.00
500µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12689096
|
Epredia
MS-1149-P |
Concentrated | 1 mL |
£490.00
1mL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12659106
|
Epredia
MS-1149-R7 |
Prediluted | 7 mL |
£293.00
7mL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12679096
|
Epredia
MS-1149-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12609106
|
Epredia
MS-1149-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12619106
|
Epredia
MS-1149-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule which is glycosylated or sialylated to produce the mature species. NCAM (CD56) is reported to express on most neuroectodermal derived cell lines, tissues, and neoplasms such as retinoblastoma, medullblastoma, astrocytoma, and neuroblastoma. It is also expressed on some mesodermally derived tumors such as rhabdomyosarcoma and also on natural killer cells.Host Species: Mouse
Clone: 56C04; same as 123A8
Isotype: IgG1/κ
Species Reactivity: Human. Others not known
Epitope: Not determined
Immunogen: Membrane preparation of a small cell lung carcinoma
Molecular Weight: 180, 145 and 125 kDa
Positive Control: Neuroblastoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD56 Ab-4 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
56C04; same as 123A8 | |
Monoclonal | |
Mouse | |
Cell Adhesion | |
Membrane preparation of a small cell lung carcinoma | |
Primary |