Learn More
CD55 Monoclonal Antibody (67 (N-L063)), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD5504
Code : 50
Additional Details : Weight : 0.02100kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD55 Monoclonal antibody specifically detects CD55 in Human samples. It is validated for Flow Cytometry.Specifications
CD55 | |
Monoclonal | |
PE | |
CD55 | |
CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC | |
Mouse | |
purified | |
ASR, ASR/IVD | |
1604 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
67 (N-L063) | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P08174 | |
CD55 | |
Human CD55 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |