missing translation for 'onlineSavingsMsg'
Learn More

CD55, FITC, clone: 143-30, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0559-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586416

  • £64.00 / 25 tests

Estimated Shipment
22-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 143-30 monoclonal antibody reacts with human CD55, the 64-73kDa molecule also known as DAF (Decay-Accelerating Factor). CD55 is a GPI-anchored single chain glycoprotein expressed on all cells in contact with serum including all hematopoietic cells and vascular endothelium; a soluble form of CD55 is also present in plasma and body fluids. CD55 is thought to play a role in the protection of cells from the damage by autologous complement via prevention of the amplification steps of the complement cascade. Antibodies against CD55 are mitogenic in the presence of phorbol esters.Applications Reported: The 143-30 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 143-30 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD55
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08174
RP11-357P18.1, CR, CROM, DAF, TC
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
143-30
FITC
CD55
Liquid
CD55
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1604
Videos
Documents

For Research Use Only.