missing translation for 'onlineSavingsMsg'
Learn More

CD55, FITC, clone: 143-30, eBioscience™

Mouse Monoclonal Antibody

£64.00 - £126.00

Specifications

Antigen CD55
Clone 143-30
Target Species Human
Conjugate FITC
Isotype IgG1, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15586416 View Documents Affymetrix eBioscience
11-0559-41
25 Tests £64.00
25 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-04-2020
Please to purchase this item. Need a web account? Register with us today!
15596416 View Documents Affymetrix eBioscience
11-0559-42
100 Tests £126.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 143-30 monoclonal antibody reacts with human CD55, the 64-73kDa molecule also known as DAF (Decay-Accelerating Factor). CD55 is a GPI-anchored single chain glycoprotein expressed on all cells in contact with serum including all hematopoietic cells and vascular endothelium; a soluble form of CD55 is also present in plasma and body fluids. CD55 is thought to play a role in the protection of cells from the damage by autologous complement via prevention of the amplification steps of the complement cascade. Antibodies against CD55 are mitogenic in the presence of phorbol esters.Applications Reported: The 143-30 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 143-30 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD55
Human
IgG1, kappa, kappa
Monoclonal
Liquid
Antibody
Store at 2-8°C. Do not freeze. Light-sensitive material.
143-30
FITC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.