missing translation for 'onlineSavingsMsg'
Learn More

CD54 (ICAM-1), PE, clone: eBioKAT-1 (KAT-1, KAT1), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0542-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15248569

  • £62.06 / 50µg


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioKAT-1 monoclonal antibody reacts with mouse CD54 (ICAM-1), which is a 95kDa member of the immunoglobulin superfamily. CD54 is expressed at low levels on leukocytes and high endothelial venules, and expression increases in response to inflammatory cytokines. ICAM-1 binds to LFA-1 and this interaction is required for the transendothelial migration of T cells. ICAM-1-deficient mice are viable, however the migration of leukocytes to sites of inflammation is reduced leading to impaired immune and inflammatory responses. Based on the regulated expression of ICAM-1, it has been suggested that ICAM-1 may increase leukocyte extravasation at sites of inflammation, whereas the constitutively high expression of ICAM-2 mediates leukocyte traffic into non-inflamed tissue. The eBioKAT-1 monoclonal antibody recognizes a different epitope than the YN1/1.7.4 monoclonal antibody.Applications Reported: This eBioKAT-1 (KAT-1, KAT1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioKAT-1 (KAT-1, KAT1) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD54 (ICAM-1)
KAT1)
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Intercellular adhesion molecule-1
IgG2a
RUO
Primary
Murine
Videos