missing translation for 'onlineSavingsMsg'
Learn More

CD54 (ICAM-1), clone: YN1/1.7.4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0541-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15277017

  • £30.70 / 50µg


Estimated Shipment
23-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The YN1/1.7.4 monoclonal antibody reacts with mouse CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes, dendritic cells, and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligands CD11a and CD11b is important in the inflammatory response. The YN1/1.7.4 monoclonal antibody recognizes a different epitope than the eBioKAT-1 monclonal antibody.Applications Reported: The YN1/1.7.4 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. YN1/1.7.4 has also been reported in blocking of CD54 in functional studies. (Please use Functional Grade purified YN1/1.7.4, cat. 16-0541, in functional assays.).Applications Tested: The YN1/1.7.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD54 (ICAM-1)
YN1/1.7.4
Unconjugated
Purified
Rat
50μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Intercellular adhesion molecule-1, ICAM1
IgG2b
RUO
Primary
Murine
Videos