Learn More
CD54 (ICAM-1) Monoclonal Antibody (YN1/1.7.4), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£55.25 - £215.00
Specifications
Antigen | CD54 (ICAM-1) |
---|---|
Clone | YN1/1.7.4 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15269599
|
Affymetrix eBioscience
13-0541-82 |
100 μg |
£116.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15279599
|
Affymetrix eBioscience
13-0541-85 |
500 μg |
£215.00
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15259599
|
Affymetrix eBioscience
13-0541-81 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The YN1/1.7.4 monoclonal antibody reacts with mouse CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110 kDa transmembrane glycoprotein expressed by monocytes, lymphocytes, dendritic cells, and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligands CD11a and CD11b is important in the inflammatory response. The YN1/1.7.4 monoclonal antibody recognizes a different epitope than the eBioKAT-1 monclonal antibody. Applications Reported: The YN1/1.7.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: The YN1/1.7.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.Specifications
CD54 (ICAM-1) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P13597 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
YN1/1.7.4 | |
Flow Cytometry | |
Biotin | |
Rat | |
Mouse | |
15894 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.