missing translation for 'onlineSavingsMsg'
Learn More

CD54 (ICAM-1), Biotin, clone: HA58, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0549-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289599

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: The HA58 monoclonal antibody reacts with human CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligand CD11a is important in the inflammatory response.Applications Reported: The HA58 antibody has been reported for use in flow cytometric analysis.Applications Tested: The HA58 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD54 (ICAM-1)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P05362
BB2, P3.58, ICAM-1, cell surface glycoprotein P3.58, intercellular adhesion molecule 1, human rhinovirus receptor, major group rhinovirus receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HA58
Biotin
ICAM1
Conjugated
ICAM1
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
3383
Videos

For Research Use Only.