missing translation for 'onlineSavingsMsg'
Learn More

CD54 (ICAM-1), APC, clone: HA58, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0549-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15557946

  • £180.80 / 100 tests


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HA58 monoclonal antibody reacts with human CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110kDa transmembrane glycoprotein expressed by monocytes, lymphocytes and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligand CD11a is important in the inflammatory response.Applications Reported: This HA58 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HA58 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD54
HA58
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Intercellular adhesion molecule-1
IgG1
RUO
Primary
Human
Videos