missing translation for 'onlineSavingsMsg'
Learn More

CD51/CD61 (Integrin alpha v beta 3), Biotin, clone: 23C6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0519-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239599

  • £49.99 / 25µg


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The 23C6 monoclonal antibody reacts with the human CD51/CD61 dimer, also known as the integrin alphav/beta3. CD51, an ∽120kDa surface molecule can also non-covalently associate with other beta subunits of the integrin family including beta1 (CD29), beta5 and beta6 to form receptors for extracellular matrix components. Heterodimers of CD51/CD61 are expressed by melanoma cells, endothelial cells and osteoclasts and at very low levels by platelets. The CD51/CD61 complex mediates adhesion to fibrinogen, fibronectin, vitronectin and thrombospondin.Applications Reported: This 23C6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 23C6 antibody has been tested by flow cytometric analysis of peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD51/CD61 (Integrin alpha v beta 3)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05106, P06756
ITGAV, ITGB3, CD51, MSK8, VNRA, VTNR, integrin alpha-V, integrin alphaVbeta3, antigen CD51, vitronectin receptor subunit alpha, CD61, GP3A, GPIIIa
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
23C6
Biotin
ITGAV
Conjugated
ITGAV, ITGB3
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
3685, 3690
Videos

For Research Use Only.