missing translation for 'onlineSavingsMsg'
Learn More

CD5, FITC, clone: 53-7.3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0051-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15238009

  • £159.81 / 500µg


EU Stock 6
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 53-7.3 monoclonal antibody reacts with mouse CD5, a 67kDa protein expressed by a majority of thymocytes, mature T cells and a subset of B cells. The expression of CD5 by a small subset of B cells characterizes a developmentally and functionally distinct lineage of B cells called B-1 cells. CD5 is a counter-receptor for CD72 and plays a role in the T-B cell interaction.Applications Reported: The 53-7.3 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 53-7.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD5
53-7.3
FITC
Conjugated
Rat
500μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Ly-1, Ly-12, Ly-A, Lyt-1
IgG2a
RUO
Primary
Murine
Videos