missing translation for 'onlineSavingsMsg'
Learn More

CD5, PE, clone: YKIX322.3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5050-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516996

  • £156.06 / 100 tests


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This YKIX322.3 monoclonal antibody reacts with canine CD5, also known as Ly-1. CD5 is a 67-kDa type I transmembrane glycoprotein expressed on thymocytes and mature T cells. Studies have demonstrated that NK cells in dogs also express CD5, but at lower levels. In dogs, CD5 is not detectable on B cells or B cell lines.Applications Reported: This YKIX322.3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This YKIX322.3 antibody has been pre-titrated and tested by flow cytometric analysis of normal canine peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD5
YKIX322.3
PE
Conjugated
IgG2a
RUO
Primary
Canine
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos