missing translation for 'onlineSavingsMsg'
Learn More

CD5 Monoclonal Antibody (UCHT2), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£115.00 - £249.00

Specifications

Antigen CD5
Clone UCHT2
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15588336
View Documents Promotion Details
Affymetrix eBioscience
25-0059-42
100 Tests
£249.00
100 tests
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15578336
View Documents Promotion Details
Affymetrix eBioscience
25-0059-41
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Description: The UCHT2 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation.\\\\The monoclonal antibody UCHT2 recognizes primate CD5. Applications Reported: This UCHT2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UCHT2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD5
5 μL/Test
Monoclonal
Liquid
RUO
P06127
IgG1 κ
Affinity chromatography
Antibody
UCHT2
Flow Cytometry
PE-Cyanine7
Mouse
Human
921
Primary
4° C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.