missing translation for 'onlineSavingsMsg'
Learn More

CD5, APC, clone: L17F12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0058-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15507866

  • £68.39 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: The L17F12 monoclonal antibody reacts with human CD5, a 67kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation.Applications Reported: This L17F12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This L17F12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD5
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P06127
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
L17F12
APC
CD5
Conjugated
CD5
IgG2a
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
921
Videos

For Research Use Only.