missing translation for 'onlineSavingsMsg'
Learn More

CD49f (Integrin alpha 6), clone: eBioGoH3 (GoH3), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0495-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227007

  • £49.78 / 50µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBioGoH3 monoclonal antibody reacts with mouse and human CD49f, also known as integrin alpha 6, very late activation antigen 6 (VLA-6 alpha chain), and platelet gpIc. CD49f is a 120 kD transmembrane protein. CD49f associates with CD29, the integrin beta 1 chain, to form the VLA-6 complex; CD49f also associates with CD104, the integrin beta 4 chain, to form the alpha 6 beta 4 complex. CD49f is expressed primarily on T cells, monocytes, platelets, epithelial and endothelial cells. CD49f expression has also been found on germinal center B cells. The eBioGoH3 antibody is cross-reactive to integrin alpha 6 on human, mouse and bovine cells. This antibody has also been reported to have functional activity in blocking the binding of integrin alpha 6 to laminin.Applications Reported: This eBioGoH3 (GoH3) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections.Applications Tested: This eBioGoH3 (GoH3) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49f (Integrin alpha 6)
eBioGoH3 (GoH3)
Unconjugated
Purified
Rat
50μg
Store at 2-8°C.
Monoclonal
Immunoprecipitation
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Integrin a6, ITGA6, VLA6
IgG2a
RUO
Primary
Murine
Videos