missing translation for 'onlineSavingsMsg'
Learn More

CD49e (Integrin alpha 5) Mouse anti-Human, PE, Clone: P1D6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-4900-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15586986

  • £78.00 / 25 tests

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Specifications

Specifications

CD49e (Integrin alpha 5)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08648
FNRA, CD49e, VLA5A, ITGA5
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
P1D6
PE
ITGA5
Liquid
ITGA5
IgG3
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3678
Videos
Documents

For Research Use Only.