Learn More
CD49e (Integrin alpha 5) Monoclonal Antibody (P1D6), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-4900-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This P1D6 monoclonal antibody reacts with human CD49e, which is also known as integrin alpha 5. Integrins constitute a family of cell surface receptors that mediate adhesion between cells or between a cell and the extracellular matrix. These proteins are heterodimers consisting of two distinct subunits, alpha and beta. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, as well as early and activated B cells. Integrins are also expressed on non-hematopoietic and cancer cells. Integrin-mediated adhesion is regulated by changes in ligand affinity (or activation), cell shape, and integrin clustering and diffusion. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to produce disulfide-linked light and heavy chains that associate with the common beta 1 subunit to form the fibronectin receptor. Studies have demonstrated the involvement of this integrin in cell adhesion, signaling, and apoptosis. The P1D6 monoclonal antibody has been reported to have blocking activity. Crossblocking studies indicate that P1D6 and eBioSAM-1 recognize a similar epitope. Applications Reported: This P1D6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This P1D6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test.
The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.Specifications
CD49e (Integrin alpha 5) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P08648 | |
Itga5 | |
Affinity chromatography | |
RUO | |
3678 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
P1D6 | |
PE | |
Itga5 | |
2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG3 |
For Research Use Only.