Learn More
CD49e (Integrin alpha 5) Monoclonal Antibody (eBioSAM-1 (SAM-1, SAM1)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£106.00 - £247.00
Specifications
Antigen | CD49e (Integrin alpha 5) |
---|---|
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570187
|
Affymetrix eBioscience
53-0496-42 |
100 Tests |
£247.00
100 tests |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15560187
|
Affymetrix eBioscience
53-0496-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBioSAM-1 monoclonal antibody reacts with human integrin alpha 5, also known as fibronectin receptor alpha chain, very late activation antigen 5 alpha, and CD49e. Integrins are composed of an alpha chain and a beta chain, which non-covalently associate to form the functional integrin. Integrin heterodimers participate in cell surface-mediated signaling and adhesion functions. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to yield disulfide linked light and heavy chains that join with Integrin beta 1 (CD29) to form the fibronectin receptor, also known as the very late activation antigen-5 (VLA-5) complex. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated B cells. Applications Reported: This eBioSAM-1 (SAM-1, SAM1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioSAM-1 (SAM-1, SAM1) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD49e (Integrin alpha 5) Monoclonal antibody specifically detects CD49e (Integrin alpha 5) in Human samples. It is validated for Flow Cytometry.Specifications
CD49e (Integrin alpha 5) | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
3678 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P08648 | |
IgG2b | |
Affinity chromatography | |
Antibody |
For Research Use Only.