missing translation for 'onlineSavingsMsg'
Learn More

CD49e (Integrin alpha 5), FITC, clone: eBioHMa5-1 (HMa5-1), eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0493-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15298109

  • £55.89 / 50µg


EU Stock 2
Add to basket

Description

Description

Description: The eBioHMalpha5-1 monoclonal antibody reacts with mouse and rat CD49e, also known as integrin alpha5 or VLA-5A. CD49e is a member of the integrin alpha chain family, and binds with CD29 (integrin beta1) to form the receptor VLA-5, which binds the ligands fibronectin and fibrinogen. CD49e is a type I, single-pass membrane protein, with a molecular weight of 135 kDa, and is expressed on multiple cell types including thymocytes, mast cells, activated T cells and splenic B cells. In addition to its role in adhesion, VLA-5 contributes to T-cell co-stimulation. The eBioHMalpha5-1 monoclonal antibody has been demonstrated to interfere with VLA-5-mediated cell adhesion, and with the degranulation of IgE-sensitized rat RBL cells stimulated on fibronectin-coated plates. Furthermore, HMalpha5-1 injected into mice subcutaneously, along with anti-CD49d and anti-CD61, is able to inhibit passive cutaneous anaphylaxis.Applications Reported: This eBioHMa5-1 (HMa5-1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioHMa5-1 (HMa5-1) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49e (Integrin alpha 5)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11688
FNRA, CD49e, VLA5A, ITGA5
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
eBioHMa5-1 (HMa5-1)
FITC
Itga5
Conjugated
Itga5
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16402
Videos

For Research Use Only.