missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA4 Monoclonal Antibody (7.2), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15433
Code : NEW
Additional Details : Weight : 0.00280kg
Description
ITGA4 Monoclonal antibody specifically detects ITGA4 in Human samples. It is validated for Flow Cytometry.Specifications
ITGA4, CD49d | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P13612 | |
ITGA4 | |
Human CD49d | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
7.2 | |
FITC | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Mouse | |
purified | |
RUO | |
3676 | |
4° C, store in dark | |
Liquid |