missing translation for 'onlineSavingsMsg'
Learn More

CD49d (Integrin alpha 4), FITC, clone: R1-2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0492-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15288109

  • £93.40 / 100µg


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Description

Description

Description: The R1-2 monoclonal antibody reacts with mouse CD49d, the 150kDa integrin alpha4 subunit. The complex of CD49d non-covalently associated with integrin beta1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin b7 and binds to the Mucosal vascular Addressin (MAdCAM-1).Applications Reported: The R1-2 antibody has been reported for use in flow cytometric analysis.Applications Tested: The R1-2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49d
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q00651
269C wild type, CD49 antigen-like family member D, VLA-4 subunit alpha, antigen CD49D, alpha-4 subunit of VLA-4 receptor, ITGA4, ITGA-IV
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
R1-2
FITC
Itga4
Conjugated
Itga4
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16401
Videos

For Research Use Only.