missing translation for 'onlineSavingsMsg'
Learn More

CD49d (Integrin alpha 4), APC, clone: MRa4-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1490-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15558016

  • £157.52 / 100µg


Estimated Shipment date
from Supplier 25-07-2018
Add to basket

Description

Description

Description: The MRa4-1 monoclonal antibody reacts with CD49d, which is also known as the integrin alpha 4 subunit. This 150-kDa cell surface protein is expressed by thymocytes, peripheral lymphocytes, monocytes, and eosinophils in complex with either integrin beta1 or integrin beta7. Outside of the immune system, CD49d is also expressed on mast cells, neural crest cells, retinal cells, and dorsal root ganglion neurons. Integrins are responsible for lymphocyte homing to Peyer's patches and mesenteric lymph nodes, as well as neural development and regeneration. Several ligands have been reported for the integrin a4b1 complex, including VCAM-1 (CD106) and fibronectin.The MRa1-4 antibody has been reported to inhibit fibronectin binding and lymphocyte migration.Applications Reported: This MRa4-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MRa4-1 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49d (Integrin alpha 4)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Itga4
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
311144
Flow Cytometry
MRa4-1
APC
Itga4
269C wild type, CD49 antigen-like family member D, VLA-4 subunit alpha, antigen CD49D, alpha-4 subunit of VLA-4 receptor, ITGA4, ITGA-IV
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Videos

For Research Use Only.