Learn More
CD49d (Integrin alpha 4) Monoclonal Antibody (9F10), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-0499-85
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The 9F10 monoclonal antibody reacts with human CD49d, the 150 kDa integrin alpha4 subunit. The complex of CD49d non-covalently associated with integrin beta1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin beta7 and binds to the Mucosal Addressin Cell-Adhesion Molecule-1 (MadCAM-1). Applications Reported: The 9F10 antibody has been reported for use in flow cytometric analysis. 9F10 has also been reported in in vitro functional assays. Applications Tested: The 9F10 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.Specifications
CD49d (Integrin alpha 4) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P13612 | |
ITGA4 | |
Affinity chromatography | |
RUO | |
3676 | |
4° C | |
Liquid |
Flow Cytometry, Functional Assay | |
9F10 | |
Functional Grade | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Mouse | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.