missing translation for 'onlineSavingsMsg'
Learn More

CD49c (Integrin alpha 3), APC, clone: P1B5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0494-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15507946

  • £81.61 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: This P1B5 monoclonal antibody reacts with human CD49c (also known as Integrin alpha 3). Integrins constitute a family of cell surface receptors that mediate adhesion between cells or between a cell and the extracellular matrix. These proteins are heterodimers consisting of two distinct subunits, alpha and beta. Integrins are expressed on hematopoietic, non-hematopoietic, and cancer cells. Integrin-mediated adhesion is regulated by changes in ligand affinity (or activation), cell shape, and integrin clustering and diffusion. Integrin alpha 3 forms a complex with the common beta 1 subunit to form the alpha3beta1 integrin. Studies have demonstrated the involvement of this integrin in wound healing and epithelial migration.The P1B5 monoclonal antibody has been reported to affect adhesion to type I and VI collagens, as well as fibronectin and laminin.Applications Reported: This P1B5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This P1B5 antibody has been pre-titrated and tested by flow cytometric analysis of HeLa cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49c (Integrin alpha 3)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P26006
CD49C, FLJ34631, FLJ34704, GAP-B3, GAPB3, MSK18, VCA-2, VL3A, VLA3a, ITGA3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
P1B5
APC
ITGA3
Conjugated
ITGA3
IgG1
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3675
Videos

For Research Use Only.