missing translation for 'onlineSavingsMsg'
Learn More

CD49b (Integrin alpha 2), FITC, clone: DX5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5971-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15288309

  • £101.39 / 100µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.Applications Reported: The DX5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The DX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49b
DX5
FITC
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ITGA2, DX5, pan-NK
IgM
RUO
Primary
Murine
Videos