missing translation for 'onlineSavingsMsg'
Learn More

CD49b (Integrin alpha 2), APC, clone: DX5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5971-63

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15320810

  • £30.74 / 10µg


Estimated Shipment
30-05-2018
Add to basket

Description

Description

Description: The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.Applications Reported: The DX5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The DX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD49b (Integrin alpha 2)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q62469
BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
DX5
APC
Itga2
Conjugated
Itga2
IgM, kappa
10μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16398
Videos

For Research Use Only.