missing translation for 'onlineSavingsMsg'
Learn More

CD49b (Integrin alpha 2), Functional Grade, clone: HMa2, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0491-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268467

  • £187.71 / 500µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HMalpha2 monoclonal antibody reacts with mouse CD49b, the 150kDa integrin alpha2 subunit. The complex of CD49b non-covalently associated with integrin beta1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by some CD4^+ T cells, IEL, NK cells, platelets and epithelial cells.Applications Reported: The HM alpha2 antibody has been reported for use in flow cytometric analysis. It has also been reported for use in in vitro functional assays.Applications Tested: The HM alpha2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD49b
HMa2
Unconjugated
Purified
Armenian Hamster
500μg
Store at 2-8°C.
Monoclonal
Functional Assay
1mg/mL
aqueous buffer, no sodium azide
Integrin a2, VLA2, ITGA2, DX5
IgG
RUO
Primary
Murine
Videos