missing translation for 'onlineSavingsMsg'
Learn More

CD48, clone: eBio156-4H9 (156-4H9), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0489-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236997

  • £61.93 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The eBio156-4H9 monoclonal antibody reacts with the human CD48 antigen, also known as B cell activation marker 1, SLAM family member 2, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. CD2 is the low affinity ligand for CD48.Recent publications have reported differential expression in mice of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors. Expression of CD48 in hematopoietic cells is found on many leukocytes with little to none on granulocytes and absent on platelets and erythrocytes. Expression increases on B cells with activation.Applications Reported: This eBio156-4H9 (156-4H9) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio156-4H9 (156-4H9) antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD48
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P09326
BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48, CD48d
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio156-4H9 (156-4H9)
Unconjugated
CD48
Purified
CD48
IgG1
100μg
Store at 2-8°C.
Primary
962
Videos

For Research Use Only.