missing translation for 'onlineSavingsMsg'
Learn More

CD48, PE, clone: HM48-1, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0481-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218559

  • £68.96 / 50µg


EU Stock 1
Add to basket

Description

Description

Description: The HM48-1 monoclonal antibody reacts with the mouse CD48 antigen; also known as BCM1, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. Recent publications have reported differential expression of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors based on the expression of these SLAM family members. Hematopoietic stem cells (HSC) are highly purified as CD150(+)CD244(-)CD48(-) cells while non-self-renewing multipotent hematopoietic progenitors (MPP) are CD244(+)CD150(-)CD48(-) and the most restricted progenitors are CD48(+)CD244(+)CD150(-). CD48 plays a critical role in adhesion and T cell activation. In the mouse, the primary counter-receptors for CD48 are CD2 and CD244. HM48-1 is reported to modulate in vitro and in vivo CD48 functions including blocking the CD48/CD2 and CD48/CD244 interactions, inhibiting the proliferative response of mitogen-activated spleen cells, providing a costimulation signal for T cells activated in vitro through their TCR, and prolonging cardiac allograft survival in vivo.Applications Reported: This HM48-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HM48-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD48
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P18181
BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48, CD48d
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
HM48-1
PE
Cd48
Purified
Cd48
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12506
Videos

For Research Use Only.